missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC356230010

 View more versions of this product

Catalog No. AC356230010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
393.55
White to light yellow
15, 1469
96.0
Authentic
213697-53-1
C26H36NP
2-Dicyclohexylphosphino-2'-(N, N-dimethylamino)biphenyl
Crystals or Crystalline Powder
1g
116° to 121°C
100.0
Glass bottle
MFCD02183572
97%
97%

Chemical Identifiers

213697-53-1
100.0
C26H36NP
MFCD02183572
≥96.0%
96.0
97%
393.55
2-Dicyclohexylphosphino-2'-(N, N-dimethylamino)biphenyl, 97%

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA