Please login to your online account to display your discounted pricing

2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Dicyclohexylphosphino-2'-(N, N-dimethylamino)biphenyl
Name Note 97%
CAS 213697-53-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C26H36NP
Formula Weight 393.55
MDL Number MFCD02183572
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356230050

  SDS
thermo fisher scientific
AC356230050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC356230010

  SDS
thermo fisher scientific
AC356230010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 393.55
Physical Form Crystals or Crystalline Powder
Quantity 5g
Color White to light yellow
Merck Index 15, 1469
TSCA TSCA
Melting Point 116° to 121°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material 2-Dicyclohexylphosphino-2'-(N, N-dimethylamino)biphenyl
Assay Percent Range 97%
Molecular Formula C26H36NP
Name Note 97%
MDL Number MFCD02183572
CAS 213697-53-1
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.