Please login to your online account to display your discounted pricing

2-(Dicyclohexylphosphino)-2-methylbiphenyl, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Dicyclohexylphosphino)-2';-methylbiphenyl
CAS 251320-86-2
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C25H33P
Formula Weight 364.51g/mol
MDL Number MFCD03094577
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380080050

  SDS
thermo fisher scientific
380080050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 108° to 112°C
Quantity 5g
Chemical Name or Material 2-(Dicyclohexylphosphino)-2';-methylbiphenyl
CAS 251320-86-2
CAS Min % 97.5
CAS Max % 100.0
Physical Form White crystals or crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C25H33P
Formula Weight 364.51g/mol
MDL Number MFCD03094577
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.