Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Dicyclohexylphosphino-2'-methylbiphenyl, 98%

$84.99 - $284.16

Chemical Identifiers

Chemical Name or Material 2-Dicyclohexylphosphino-2'-methylbiphenyl
CAS 251320-86-2
Molecular Formula C25H33P
Formula Weight 364.51g/mol
MDL Number MFCD03094577
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2764703

 
alfa aesar™
H27647-03
1g Each for $84.99

AAH2764706

 
alfa aesar™
H27647-06
5g Each for $284.16
Description & Specifications

Specifications

Melting Point 107° to 110°C
Quantity 1g
Chemical Name or Material 2-Dicyclohexylphosphino-2'-methylbiphenyl
CAS 251320-86-2
Assay 98%
Molecular Formula C25H33P
Formula Weight 364.51g/mol
MDL Number MFCD03094577
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.