missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC389722500

 View more versions of this product

Catalog No. AC389722500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl
251320-85-1
100.0
Glass bottle
≥97.5%
C27H37P
MFCD09744023
250mg
97.5
Authentic
Crystalline Powder
98%
392.56
White

Chemical Identifiers

251320-85-1
100.0
C27H37P
MFCD09744023
≥97.5%
97.5
98%
392.56
2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.