Please login to your online account to display your discounted pricing

2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Dicyclohexylphosphino-2', 6'-dimethoxybiphenyl
CAS 657408-07-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C26H35O2P
Formula Weight 410.54
Synonym Sphos
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC441860010

  SDS
thermo fisher scientific
AC441860010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 410.54
Quantity 1g
Color Off-white to pale brown
Melting Point 162° to 167°C
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Chemical Name or Material 2-Dicyclohexylphosphino-2', 6'-dimethoxybiphenyl
CAS 657408-07-6
Molecular Formula C26H35O2P
Elemental Analysis 73.00 to 79.10 % C 8.25 to 8.94 % H
Synonym Sphos
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.