Please login to your online account to display your discounted pricing

2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC441860050

 View more versions of this product

Catalog No. AC441860050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 410.54
Physical Form Powder
Quantity 5g
Color Off-white to pale brown
Melting Point 162° to 167°C
CAS Min % 96.0
CAS Max % 100.0
Synonym Sphos
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C26H35O2P
Chemical Name or Material 2-Dicyclohexylphosphino-2', 6'-dimethoxybiphenyl
CAS 657408-07-6
Packaging Glass bottle
Infrared Spectrum Authentic
Elemental Analysis 73.00 to 79.10 % C 8.25 to 8.94 % H
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.

Specifications

Chemical Name or Material 2-Dicyclohexylphosphino-2', 6'-dimethoxybiphenyl
CAS 657408-07-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C26H35O2P
Formula Weight 410.54
Synonym Sphos
Accessories

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.

SDS & Safety Documents