Please login to your online account to display your discounted pricing

2-(Dicyclohexylphosphino)-2',4',6'-triisopropylbiphenyl, +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Dicyclohexylphosphino)-2', 4', 6'-triisopropylbiphenyl
CAS 564483-18-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C33H49P
Formula Weight 476.73
MDL Number MFCD04117682
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387140250

  SDS
fisher scientific acros
AC387140250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 476.73
Assay ≥98.0%
Quantity 25g
Color White
Melting Point 183° to 187°C
Merck Index 15, 1469
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 2-(Dicyclohexylphosphino)-2', 4', 6'-triisopropylbiphenyl
Molecular Formula C33H49P
Assay Percent Range 98+%
MDL Number MFCD04117682
CAS 564483-18-7
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.