Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl, 98+%

$83.39 - $1,442.15

Chemical Identifiers

Chemical Name or Material 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl
CAS 564483-18-7
Molecular Formula C33H49P
Formula Weight 476.73
MDL Number MFCD04117682
Synonym X-PHOS
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2748103

 
alfa aesar™
H2748103
1g Each for $83.39

AAH2748114

 
alfa aesar™
H2748114
25g Each for $1,442.15
Description & Specifications

Specifications

Formula Weight 476.73
Chemical Name or Material 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl
Assay ≥98%
MDL Number MFCD04117682
CAS 564483-18-7
Melting Point 187° to 190°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C33H49P
Synonym X-PHOS
Health Hazard 1 Warning
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.