Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Dicyanomethylene-1,3-indanedione, 98%


Quantity 5g

Manufacturer: alfa aesar™  L0892606

 View more versions of this product

Catalog No. AAL0892606

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 208.18
MDL Number MFCD00051930
Chemical Name or Material 2-Dicyanomethylene-1,3-indanedione
Assay 98%
Molecular Formula C12H4N2O2
CAS 16954-74-8
Health Hazard 2 GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Health Hazard 1 Danger
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 5g
Melting Point ≈270°C
Beilstein 2213594
Health Hazard 3 GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material 2-Dicyanomethylene-1,3-indanedione
CAS 16954-74-8
Molecular Formula C12H4N2O2
Formula Weight 208.18
MDL Number MFCD00051930
UN Number UN3439
Beilstein 2213594
Accessories

Danger

GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.

SDS & Safety Documents