Please login to your online account to display your discounted pricing

2-(Di-tert-butylphosphino)biphenyl, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Di-tert-butylphosphino)biphenyl
Name Note 99%
CAS 224311-51-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C20H27P
Formula Weight 298.41
MDL Number MFCD01862440
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356210050

  SDS
thermo fisher scientific
AC356210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC356215000

  SDS
fisher scientific acros
356215000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystals or Crystalline Powder
Assay ≥98.5%
Formula Weight 298.41
Quantity 5g
Color White
Merck Index 15, 1469
TSCA TSCA
Melting Point 84° to 87°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2-(Di-tert-butylphosphino)biphenyl
Name Note 99%
Assay Percent Range 99%
Molecular Formula C20H27P
MDL Number MFCD01862440
CAS 224311-51-7
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.