Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl, 98%

Chemical Identifiers

Chemical Name or Material 2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl
CAS 224311-49-3
Molecular Formula C22H32NP
Formula Weight 341.47
MDL Number MFCD03426986
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27209MD

 
alfa aesar™
H27209MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 341.47
Chemical Name or Material 2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl
Assay 98%
Molecular Formula C22H32NP
MDL Number MFCD03426986
CAS 224311-49-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 114° to 117°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.