Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Di-tert-butylphosphino)-2'-methylbiphenyl, 99%

$32.49 - $91.88

Chemical Identifiers

Chemical Name or Material 2-(Di-tert-butylphosphino)-2'-methylbiphenyl
CAS 255837-19-5
Molecular Formula C21H29P
Formula Weight 312.43g/mol
MDL Number MFCD03453047
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2774203

 
alfa aesar™
H27742-03
1g Each for $91.88

AAH27742MD

 
alfa aesar™
H27742-MD
250mg Each for $32.49
Description & Specifications

Specifications

Melting Point 90° to 93°C
Quantity 1g
Chemical Name or Material 2-(Di-tert-butylphosphino)-2'-methylbiphenyl
CAS 255837-19-5
Assay 99%
Molecular Formula C21H29P
Formula Weight 312.43g/mol
MDL Number MFCD03453047
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.