missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, 97%, (patented by MIT Cambri, ACROS Organics™
Chemical Identifiers
CAS | 564483-19-8 |
---|---|
Molecular Formula | C29H45P |
Molecular Weight (g/mol) | 424.66 |
MDL Number | MFCD06411306 |
InChI Key | SACNIGZYDTUHKB-UHFFFAOYSA-N |
Synonym | 2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-di-t-butylphosphino-2',4',6'-triisopropylbiphenyl, 2-di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, di-t-bu-xphos, di-tert-butyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, di-tert-butyl 2',4',6'-triisopropylbiphenyl-2-yl phosphine, t-bu xphos, t-butylxphos, tbuxphos, tert-butyl xphos |
PubChem CID | 11618717 |
IUPAC Name | ditert-butyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane |
SMILES | CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C)C(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC429840050
|
Thermo Fisher Scientific
AC429840050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
564483-19-8 | |
424.66 | |
SACNIGZYDTUHKB-UHFFFAOYSA-N | |
11618717 | |
CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C)C(C)C |
C29H45P | |
MFCD06411306 | |
2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-di-t-butylphosphino-2',4',6'-triisopropylbiphenyl, 2-di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, di-t-bu-xphos, di-tert-butyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, di-tert-butyl 2',4',6'-triisopropylbiphenyl-2-yl phosphine, t-bu xphos, t-butylxphos, tbuxphos, tert-butyl xphos | |
ditert-butyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane |
Specifications
79.15 to 84.90% C | |
consistent with structure | |
White | |
5g | |
96.0 | |
96% min. (HPLC) | |
MFCD06411306 | |
SACNIGZYDTUHKB-UHFFFAOYSA-N | |
ditert-butyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane | |
11618717 | |
Crystalline Powder | |
2-Di-tert-butylphosphino-2', 4', 6'-triisopropylbiphenyl |
Authentic | |
Glass bottle | |
148°C to 149°C | |
564483-19-8 | |
100.0 | |
C29H45P | |
2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-di-t-butylphosphino-2',4',6'-triisopropylbiphenyl, 2-di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, di-t-bu-xphos, di-tert-butyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, di-tert-butyl 2',4',6'-triisopropylbiphenyl-2-yl phosphine, t-bu xphos, t-butylxphos, tbuxphos, tert-butyl xphos | |
CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C)C(C)C | |
424.66 | |
424.66 | |
97% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning