Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Di-p-tolylphosphino)benzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2-(Di-p-tolylphosphino)benzaldehyde
CAS 1202865-03-9
Formula Weight 318.12
MDL Number MFCD15143661
Synonym 2-[Bis(4-methylphenyl)phosphino]benzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6318303

  SDS
alfa aesar™
H6318303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6318306

  SDS
alfa aesar™
H6318306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63183MD

  SDS
alfa aesar™
H63183MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 105° to 111°C
Formula Weight 318.12
Sensitivity Air Sensitive
Chemical Name or Material 2-(Di-p-tolylphosphino)benzaldehyde
Assay 97%
MDL Number MFCD15143661
Physical Form Solid
Synonym 2-[Bis(4-methylphenyl)phosphino]benzaldehyde
CAS 1202865-03-9
Color Yellow
Quantity 1g