missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Deoxyuridine-5'-triphosphate trisodium salt, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J64716MC

 View more versions of this product

Catalog No. AAJ64716MC



Description

Description

2'-Deoxyuridine-5'-triphosphate disodium salt is used in conjunction with uracil DNA glycosylase to prevent carryover contamination in PCR. The dUTP is substituted for dTTP in the PCR reaction, used to study the distribution, specificity and kinetics of deoxyuridine 5?-triphosphate nucleotidohydrolase(s) (dUTPase, EC 3.6.1.23). dUTPase are important enzymes that protect cells from the incorporation of dUTP into DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyuridine-5′-triphosphate disodium salt is used in conjunction with uracil DNA glycosylase to prevent carryover contamination in PCR. The dUTP is substituted for dTTP in the PCR reaction, used to study the distribution, specificity and kinetics of deoxyuridine 5′-triphosphate nucleotidohydrolase(s) (dUTPase, EC 3.6.1.23). dUTPase are important enzymes that protect cells from the incorporation of dUTP into DNA.

Solubility
Soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

2'-Deoxyuridine-5'-triphosphate disodium salt
White
MFCD00084701
2'-deoxyuridine-5'-triphosphate trisodium salt
AYFBRVJQNMGUSV-MILVPLDLSA-N
[[(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
131843218
95%
102814-08-4
C9H15N2Na3O14P3
100 mg
Soluble in water.
C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O.[Na].[Na].[Na]
537.11
537.11
Powder

Chemical Identifiers

102814-08-4
537.11
AYFBRVJQNMGUSV-MILVPLDLSA-N
131843218
C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O.[Na].[Na].[Na]
C9H15N2Na3O14P3
MFCD00084701
2'-deoxyuridine-5'-triphosphate trisodium salt
[[(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only