missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Deoxyinosine, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 226361000

 View more versions of this product

Catalog No. AC226361000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2'-Deoxyinosine
Glass bottle
White
100mg
890-38-0
100.0
C10H12N4O4
2'-deoxyinosine, 2-deoxyinosine, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, d-ino, delta-ino, deoxyinosine, hn0rq6sbwq, inosine, 2'-deoxy, unii-hn0rq6sbwq
C1C(C(OC1N2C=NC3=C2NC=NC3=O)CO)O
252.23
CHEBI:28997
Powder
Authentic
+ 21.00
>250.0°C
99%
98.0
98% min. (HPLC)
MFCD00005762
VGONTNSXDCQUGY-RRKCRQDMSA-N
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
65058
252.23
98+%

Chemical Identifiers

890-38-0
252.23
VGONTNSXDCQUGY-RRKCRQDMSA-N
65058
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H12N4O4
MFCD00005762
2'-deoxyinosine, 2-deoxyinosine, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, d-ino, delta-ino, deoxyinosine, hn0rq6sbwq, inosine, 2'-deoxy, unii-hn0rq6sbwq
CHEBI:28997
C1C(C(OC1N2C=NC3=C2NC=NC3=O)CO)O
Safety and Handling

Safety and Handling

EINECSNumber : 212-964-1

TSCA : TSCA

SDS Documents