missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Deoxyinosine, 98+%, ACROS Organics™

Chemical Identifiers

CAS 890-38-0
Molecular Formula C10H12N4O4
Molecular Weight (g/mol) 252.23
MDL Number MFCD00005762
InChI Key VGONTNSXDCQUGY-RRKCRQDMSA-N
Synonym 2'-deoxyinosine, 2-deoxyinosine, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, d-ino, delta-ino, deoxyinosine, hn0rq6sbwq, inosine, 2'-deoxy, unii-hn0rq6sbwq
PubChem CID 65058
ChEBI CHEBI:28997
IUPAC Name 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES C1C(C(OC1N2C=NC3=C2NC=NC3=O)CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC226361000
SDS View Documents
Thermo Fisher Scientific
226361000
100mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC226365000
SDS View Documents
Thermo Fisher Scientific
AC226365000
500mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

890-38-0
252.23
VGONTNSXDCQUGY-RRKCRQDMSA-N
65058
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H12N4O4
MFCD00005762
2'-deoxyinosine, 2-deoxyinosine, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, d-ino, delta-ino, deoxyinosine, hn0rq6sbwq, inosine, 2'-deoxy, unii-hn0rq6sbwq
CHEBI:28997
C1C(C(OC1N2C=NC3=C2NC=NC3=O)CO)O

Specifications

Authentic
+ 21.00
>250.0°C
890-38-0
100.0
C10H12N4O4
2'-deoxyinosine, 2-deoxyinosine, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, d-ino, delta-ino, deoxyinosine, hn0rq6sbwq, inosine, 2'-deoxy, unii-hn0rq6sbwq
C1C(C(OC1N2C=NC3=C2NC=NC3=O)CO)O
252.23
CHEBI:28997
Powder
2'-Deoxyinosine, 99%
Glass bottle
White
100mg
98.0
98% min. (HPLC)
MFCD00005762
VGONTNSXDCQUGY-RRKCRQDMSA-N
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
65058
252.23
98+%
Safety and Handling

Safety and Handling

EINECSNumber : 212-964-1

TSCA : TSCA

SDS