missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Deoxyinosine, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC226365000
Specifications
2'-Deoxyinosine | |
Glass bottle | |
+ 21.00 | |
>250.0°C | |
99% | |
98.0 | |
98% min. (HPLC) | |
MFCD00005762 | |
VGONTNSXDCQUGY-RRKCRQDMSA-N | |
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
65058 | |
252.23 | |
98+% |
Authentic | |
+21° (22°C c=1,H2O) | |
White | |
500mg | |
890-38-0 | |
100.0 | |
C10H12N4O4 | |
2'-deoxyinosine, 2-deoxyinosine, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, d-ino, delta-ino, deoxyinosine, hn0rq6sbwq, inosine, 2'-deoxy, unii-hn0rq6sbwq | |
C1C(C(OC1N2C=NC3=C2NC=NC3=O)CO)O | |
252.23 | |
CHEBI:28997 | |
Powder |
Chemical Identifiers
890-38-0 | |
252.23 | |
VGONTNSXDCQUGY-RRKCRQDMSA-N | |
65058 | |
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
C10H12N4O4 | |
MFCD00005762 | |
2'-deoxyinosine, 2-deoxyinosine, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, d-ino, delta-ino, deoxyinosine, hn0rq6sbwq, inosine, 2'-deoxy, unii-hn0rq6sbwq | |
CHEBI:28997 | |
C1C(C(OC1N2C=NC3=C2NC=NC3=O)CO)O |
Safety and Handling
EINECSNumber : 212-964-1
TSCA : TSCA