missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Deoxyguanosine hydrate, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 226730250

 View more versions of this product

Catalog No. AC226730250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1.12 to 1.18 (A250/A260) 0.65 to 0.69 (A280/A260)
(on dry substance) :
Authentic
98% min. (lambda max. 252.5nm,pH 7) molar absorptivity 13700+/-4%,on dry basis
8% Max. (K.F.)
300°C
99+%
99.0
99% min. (HPLC)
MFCD00150760
YKBGVTZYEHREMT-KVQBGUIXSA-N
2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
187790
267.12
99+%
2′-Deoxyguanosine hydrate
300°C
Glass bottle
0.1% max.
White
25mg
961-07-9
100.0
C10H13N5O4·xH2O
2'-deoxy-guanosine, 2'-deoxyguanosine, 2-deoxyguanosine, 2;-deoxyguanosine, deoxyguanosine, guanine deoxy nucleoside, guanine deoxyriboside, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl, guanosine, 2'-deoxy, unii-g9481n71ro
C1C(C(OC1N2C=NC3=C2NC(=NC3=O)N)CO)O
267.12
CHEBI:17172
Powder

Chemical Identifiers

961-07-9
267.12
YKBGVTZYEHREMT-KVQBGUIXSA-N
187790
2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H13N5O4·xH2O
MFCD00150760
2'-deoxy-guanosine, 2'-deoxyguanosine, 2-deoxyguanosine, 2;-deoxyguanosine, deoxyguanosine, guanine deoxy nucleoside, guanine deoxyriboside, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl, guanosine, 2'-deoxy, unii-g9481n71ro
CHEBI:17172
C1C(C(OC1N2C=NC3=C2NC(=NC3=O)N)CO)O
Safety and Handling

Safety and Handling

EINECSNumber : 213-505-8

RTECSNumber : MF8760000

TSCA : TSCA

SDS Documents