missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Deoxycytidine-5'-triphosphate disodium salt

$62.92 - $288.38

Chemical Identifiers

CAS 102783-51-7
Molecular Formula C18H30N6Na2O26P6
Molecular Weight (g/mol) 978.276
MDL Number MFCD00084683
InChI Key BQFLCFZDELHQIZ-NGLQWXHTSA-N
Synonym 5'-dCTP Na2; 2'-Deoxy-5'-triphosphocytidine disodium salt
PubChem CID 131636617
IUPAC Name disodium;(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-2-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolan-3-olate
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-].C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65686MB View Documents Alfa Aesar™
J65686MB
25mg Each for $62.92
 
AAJ65686MD View Documents Alfa Aesar™
J65686MD
250mg Each for $288.38
 
Description

Description

Applications
2′-Deoxycytidine 5′-triphosphate disodium salt is a nucleoside triphosphatase used for DNA synthesis with DNA polymerases and reverse transcriptases. 2′-Deoxycytidine 5′-triphosphate disodium salt is one of the nucleoside triphosphates used for DNA polymerase driven reactions such as polymerase chain reaction, DNA sequencing and other molecular biology methods.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Specifications

Chemical Identifiers

102783-51-7
978.276
BQFLCFZDELHQIZ-NGLQWXHTSA-N
131636617
C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-].C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-].[Na+].[Na+]
C18H30N6Na2O26P6
MFCD00084683
5'-dCTP Na2; 2'-Deoxy-5'-triphosphocytidine disodium salt
disodium;(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-2-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolan-3-olate

Specifications

White
102783-51-7
MFCD00084683
BQFLCFZDELHQIZ-NGLQWXHTSA-N
disodium;(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-2-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolan-3-olate
131636617
Powder
25mg
C18H30N6Na2O26P6
5'-dCTP Na2; 2'-Deoxy-5'-triphosphocytidine disodium salt
C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-].C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-].[Na+].[Na+]
978.276
511.12
2'-Deoxycytidine-5'-triphosphate disodium salt
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C