Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Deoxycytidine-5'-diphosphate trisodium salt, 96%

$164.24 - $164.24

Chemical Identifiers

Chemical Name or Material 2'-Deoxycytidine-5'-diphosphate trisodium salt
CAS 151151-32-5
Molecular Formula C9H12N3Na3O10P2
Formula Weight 453.12
MDL Number MFCD00056068
UN Number UN2811
Synonym dCDP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65408MB

  SDS
alfa aesar™
J65408MB
25mg Each for $164.24
Description & Specifications

Specifications

Formula Weight 453.12
Chemical Name or Material 2'-Deoxycytidine-5'-diphosphate trisodium salt
Molecular Formula C9H12N3Na3O10P2
CAS 151151-32-5
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Physical Form Powder
Assay 98%
MDL Number MFCD00056068
UN Number UN2811
Synonym dCDP
Health Hazard 1 Danger
Color White
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 25mg
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2′-Deoxycytidine 5′-diphosphate (dCDP) is used as a substrate of CDP (nucleoside diphosphate) kinase to produce dCTP in support of DNA biosynthesis and reverse transcription.

Solubility
Soluble in water.

Notes
Store at -20°C. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C