missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Deoxyadenosine-5'-triphosphate disodium salt, 98%

$445.68 - $445.68

Chemical Identifiers

CAS 74299-50-6
Molecular Formula C10H14N5Na2O12P3
Molecular Weight (g/mol) 535.146
MDL Number MFCD00080335
InChI Key JEKDCIBJADJZSK-OJSHLMAWSA-L
Synonym 2'-deoxyadenosine-5'-triphosphate disodium salt
PubChem CID 131674273
IUPAC Name disodium;[[[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)(O)OP(=O)([O-])[O-])O.[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64045MC View Documents Alfa Aesar™
J64045MC
100mg Each for $445.68
 
Description

Description

Applications
2′-Deoxyadenosine-5′-triphosphate disodium salt is used as a substrate by a variety of polymerases including DNA polymerase(s) and reverse transcriptase(s).

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

74299-50-6
535.146
JEKDCIBJADJZSK-OJSHLMAWSA-L
131674273
C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)(O)OP(=O)([O-])[O-])O.[Na+].[Na+]
C10H14N5Na2O12P3
MFCD00080335
2'-deoxyadenosine-5'-triphosphate disodium salt
disodium;[[[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate

Specifications

White
74299-50-6
MFCD00080335
JEKDCIBJADJZSK-OJSHLMAWSA-L
disodium;[[[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate
131674273
Powder
2'-Deoxyadenosine-5'-triphosphate disodium salt
100mg
C10H14N5Na2O12P3
2'-deoxyadenosine-5'-triphosphate disodium salt
C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)(O)OP(=O)([O-])[O-])O.[Na+].[Na+]
535.146
535.14
98%
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C