Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Deoxyadenosine-5'-triphosphate disodium salt, 97%

Chemical Identifiers

Chemical Name or Material 2'-Deoxyadenosine-5'-triphosphate disodium salt
CAS 1927-31-7
Formula Weight 535.14
MDL Number MFCD00080335
Synonym dATP-Na2; 2'-Deoxyadenosine 5'-triphosphoric acid disodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65542ME

 
alfa aesar™
J65542ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 535.14
Physical Form Powder
Assay 97%
MDL Number MFCD00080335
CAS 1927-31-7
Synonym dATP-Na2; 2'-Deoxyadenosine 5'-triphosphoric acid disodium salt
Chemical Name or Material 2'-Deoxyadenosine-5'-triphosphate disodium salt
Storage Note 1 -30° to -10°C
Color White
Quantity 500mg
Safety and Handling

-30° to -10°C