missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Deoxyadenosine-5'-monophosphate, 98%

$60.28 - $777.23

Chemical Identifiers

CAS 653-63-4
Molecular Formula C10H14N5O6P
Molecular Weight (g/mol) 331.225
MDL Number MFCD00005753
InChI Key KHWCHTKSEGGWEX-RRKCRQDMSA-N
Synonym damp, 2'-deoxyadenosine-5'-monophosphate, 2'-deoxyadenosine 5'-monophosphate, 2'-deoxyadenylic acid, deoxyadenylic acid, deoxy-amp, 2'-deoxy-amp, 2'-deoxyadenosine 5'-phosphate, 2'-damp, deoxyadenosine monophosphate
PubChem CID 12599
ChEBI CHEBI:17713
IUPAC Name [(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6440203 View Documents Alfa Aesar™
J6440203
1g Each for $60.28
 
AAJ6440206 View Documents Alfa Aesar™
J6440206
5g Each for $239.03
 
AAJ6440214 View Documents Alfa Aesar™
J6440214
25g Each for $777.23
 
Description

Description

Solubility
Soluble in 1N ammonium hhydroxide (50 mg/ml) and water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

653-63-4
331.225
KHWCHTKSEGGWEX-RRKCRQDMSA-N
12599
[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
C10H14N5O6P
MFCD00005753
damp, 2'-deoxyadenosine-5'-monophosphate, 2'-deoxyadenosine 5'-monophosphate, 2'-deoxyadenylic acid, deoxyadenylic acid, deoxy-amp, 2'-deoxy-amp, 2'-deoxyadenosine 5'-phosphate, 2'-damp, deoxyadenosine monophosphate
CHEBI:17713
C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)O)O

Specifications

White
653-63-4
MFCD00005753
KHWCHTKSEGGWEX-RRKCRQDMSA-N
[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
12599
331.22
98%
1g
C10H14N5O6P
damp, 2'-deoxyadenosine-5'-monophosphate, 2'-deoxyadenosine 5'-monophosphate, 2'-deoxyadenylic acid, deoxyadenylic acid, deoxy-amp, 2'-deoxy-amp, 2'-deoxyadenosine 5'-phosphate, 2'-damp, deoxyadenosine monophosphate
C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)O)O
331.225
CHEBI:17713
Crystalline Powder
2'-Deoxyadenosine-5'-monophosphate
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C