missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Deoxyadenosine-5'-diphosphate trisodium salt, 98%

$727.88 - $727.88

Chemical Identifiers

CAS 72003-83-9
Molecular Formula C10H15N5Na2O9P2
Molecular Weight (g/mol) 457.183
MDL Number MFCD00083610
InChI Key PMCITFSCGIPAKY-OJSHLMAWSA-N
Synonym 2'-deoxyadenosine-5'-diphosphate disodium salt
PubChem CID 131858105
IUPAC Name [(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)(O)O)O.[Na].[Na]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65307MC View Documents Alfa Aesar™
J65307MC
100mg Each for $727.88
 
Description

Description

Applications
2′-Deoxyadenosine 5′-diphosphate is used for the synthesis of deoxyadenylate oligonucleotides with enzymes such as polynucleotide phosphorylase from Escherichia coli. dADP is used as an inhibitor of bacterial poly(A) polymerase. DeoxyADP may be use as an alternative substrate or inhibitor for studies of ATPase and DNA and RNA polymerase specificity.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

72003-83-9
457.183
PMCITFSCGIPAKY-OJSHLMAWSA-N
131858105
C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)(O)O)O.[Na].[Na]
C10H15N5Na2O9P2
MFCD00083610
2'-deoxyadenosine-5'-diphosphate disodium salt
[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium

Specifications

White
72003-83-9
MFCD00083610
PMCITFSCGIPAKY-OJSHLMAWSA-N
[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium
131858105
Powder
2'-Deoxyadenosine-5'-diphosphate trisodium salt
100mg
C10H15N5Na2O9P2
2'-deoxyadenosine-5'-diphosphate disodium salt
C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)(O)O)O.[Na].[Na]
457.183
477.15
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C