missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxy-5-fluorocytidine 98.0+%, TCI America™
$460.43 - $1585.38
Chemical Identifiers
| CAS | 10356-76-0 |
|---|---|
| Molecular Formula | C9H12FN3O4 |
| Molecular Weight (g/mol) | 245.21 |
| MDL Number | MFCD00077348 |
| InChI Key | IDYKCXHJJGMAEV-UHFFFAOYNA-N |
| PubChem CID | 515328 |
| IUPAC Name | 4-amino-5-fluoro-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one |
| SMILES | NC1=NC(=O)N(C=C1F)C1CC(O)C(CO)O1 |
Chemical Identifiers
| 10356-76-0 | |
| 245.21 | |
| IDYKCXHJJGMAEV-UHFFFAOYNA-N | |
| 4-amino-5-fluoro-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one |
| C9H12FN3O4 | |
| MFCD00077348 | |
| 515328 | |
| NC1=NC(=O)N(C=C1F)C1CC(O)C(CO)O1 |
Specifications
| 10356-76-0 | |
| White | |
| MFCD00077348 | |
| IDYKCXHJJGMAEV-UHFFFAOYNA-N | |
| 4-amino-5-fluoro-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one | |
| 515328 | |
| ≥98.0% (HPLC,T) | |
| 2′-Deoxy-5-fluorocytidine |
| 196°C | |
| C9H12FN3O4 | |
| 1 g | |
| NC1=NC(=O)N(C=C1F)C1CC(O)C(CO)O1 | |
| 245.21 | |
| 245.21 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : HA3850000
TSCA : No