missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Cyclopropyl-L-glycine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H65755MD
Description
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Store at 4°C. Incompatible with heat and oxidizing agents. Protect from heat.
Specifications
2-Cyclopropyl-L-glycine | |
White | |
MFCD06659116 | |
l-cyclopropylglycine, s-2-amino-2-cyclopropylacetic acid, h-cyclopropyl-gly-oh, 2-cyclopropyl-l-glycine, l-cyclopropyl glycine, 2s-2-amino-2-cyclopropylacetic acid, s-amino-cyclopropyl-acetic acid, alpha-cyclopropylglycine, s-amino cyclopropyl acetic acid | |
BUSBCPMSNBMUMT-BYPYZUCNSA-N | |
(2S)-2-azaniumyl-2-cyclopropylacetate | |
1501944 | |
97% | |
Crystalline |
49606-99-7 | |
C5H9NO2 | |
5728866 | |
Soluble in water. Insoluble in ethanol. | |
[NH3+][C@@H](C1CC1)C([O-])=O | |
115.13 | |
115.13 | |
250 mg |
Chemical Identifiers
49606-99-7 | |
115.13 | |
BUSBCPMSNBMUMT-BYPYZUCNSA-N | |
1501944 |
C5H9NO2 | |
MFCD06659116 | |
l-cyclopropylglycine, s-2-amino-2-cyclopropylacetic acid, h-cyclopropyl-gly-oh, 2-cyclopropyl-l-glycine, l-cyclopropyl glycine, 2s-2-amino-2-cyclopropylacetic acid, s-amino-cyclopropyl-acetic acid, alpha-cyclopropylglycine, s-amino cyclopropyl acetic acid | |
[NH3+][C@@H](C1CC1)C([O-])=O |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only