missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyclopentylphenol, +98%, ACROS Organics™

Manufacturer:  Acros Organics AC426510010

 View more versions of this product

Catalog No. AC426510010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Cyclopentylphenol
Consistent with structure (1H-NMR)
148°C to 150°C (18.0mmHg)
36°C
1518-84-9
100.0
C11H14O
2-cyclopentyl phenol
JHEKSKQMOBLXQS-UHFFFAOYSA-N
2-cyclopentylphenol
80285
Liquid
Authentic
Glass bottle
Colorless to Yellow
1g
98.0
98+%
MFCD00009952
Solubility in water: insoluble
C1CCC(C1)C2=CC=CC=C2O
162.23
162.23
≥97.5% (GC)

Chemical Identifiers

1518-84-9
162.23
JHEKSKQMOBLXQS-UHFFFAOYSA-N
80285
C1CCC(C1)C2=CC=CC=C2O
C11H14O
MFCD00009952
2-cyclopentyl phenol
2-cyclopentylphenol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Documents