Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyclopentylethanol, 98+%

$118.99 - $118.99

Chemical Identifiers

Chemical Name or Material 2-Cyclopentylethanol
CAS 766-00-7
Molecular Formula C7H14O
Formula Weight 114.19
MDL Number MFCD00019301
Beilstein 2069860
Synonym 2-(2-Hydroxyethyl)cyclopentane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2234403

 
alfa aesar™
B2234403
1g Each for $118.99
Description & Specifications

Specifications

Formula Weight 114.19
Chemical Name or Material 2-Cyclopentylethanol
Assay ≥98%
Molecular Formula C7H14O
MDL Number MFCD00019301
CAS 766-00-7
Beilstein 2069860
Synonym 2-(2-Hydroxyethyl)cyclopentane
Health Hazard 1 Warning
Flash Point 70°C (158°F)
Boiling Point 182°C
Quantity 1g
Safety and Handling

Warning