Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyclopentylcyclopentanone, 99%

$57.57 - $200.41

Chemical Identifiers

Chemical Name or Material 2-Cyclopentylcyclopentanone
CAS 4884-24-6
Molecular Formula C10H16O
Formula Weight 152.24
MDL Number MFCD00051419
Beilstein 1306869
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1250306

  SDS
alfa aesar™
L1250306
5g Each for $57.57

AAL1250314

  SDS
alfa aesar™
L1250314
25g Each for $200.41
Description & Specifications

Specifications

Formula Weight 152.24
Chemical Name or Material 2-Cyclopentylcyclopentanone
CAS 4884-24-6
MDL Number MFCD00051419
Assay 99%
Flash Point 104°C (219°F)
Beilstein 1306869
Quantity 5g
Boiling Point 230° to 233°C
TSCA TSCA
Molecular Formula C10H16O