Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyclohexyloxypyridine-3-boronic acid pinacol ester, 99%

$281.77 - $281.77

Chemical Identifiers

Chemical Name or Material 2-Cyclohexyloxypyridine-3-boronic acid pinacol ester
CAS 1073354-43-4
Molecular Formula C17H26BNO3
Formula Weight 303.21
MDL Number MFCD07781171
Synonym 2-Cyclohexyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5011103

  SDS
alfa aesar™
H5011103
1g Each for $281.77
Description & Specifications

Specifications

Formula Weight 303.21
CAS 1073354-43-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD07781171
Chemical Name or Material 2-Cyclohexyloxypyridine-3-boronic acid pinacol ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C17H26BNO3
Synonym 2-Cyclohexyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Assay 99%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.