Please login to your online account to display your discounted pricing

2-Cyclohexen-1-ol, 95%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC111230100

 View more versions of this product

Catalog No. AC111230100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 98.14
Quantity 10g
Color Clear colorless
Density 0.984g/mL
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Cyclohexen-1-ol
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C6H10O
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Packaging Glass bottle
Linear Formula C6H9OH
MDL Number MFCD00001570
CAS 822-67-3
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Cyclohexen-1-ol
CAS 822-67-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H10O
Linear Formula C6H9OH
Formula Weight 98.14
MDL Number MFCD00001570
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapour.

SDS & Safety Documents