missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyanothioacetamide, 98%

Manufacturer:  Alfa Aesar™ A1543622

 View more versions of this product

Catalog No. AAA1543622



Description

Description

Applications
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.

Solubility
Does not mix well with water.

Notes
Light Sensitive. Store in the dark.
Specifications

Specifications

2-Cyanothioacetamide
100g
C3H4N2S
UN3439
2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide
C(C#N)C(=S)N
100.139
100.14
114°C to 120°C
7357-70-2
MFCD00010025
1699934
BHPYMZQTCPRLNR-UHFFFAOYSA-N
2-cyanoethanethioamide
1416277
98%

Chemical Identifiers

7357-70-2
100.139
BHPYMZQTCPRLNR-UHFFFAOYSA-N
1416277
C(C#N)C(=S)N
C3H4N2S
MFCD00010025
2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide
2-cyanoethanethioamide
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents