Learn More
2-Cyanothiazole, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H63425MD
Description
2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2-Cyanothiazole | |
C4H2N2S | |
250 mg | |
2-cyanothiazole, thiazole-2-carbonitrile, 2-thiazolecarbonitrile, 2-cyanothiazol | |
ZIRGWUZHKJDYKT-UHFFFAOYSA-N | |
1,3-thiazole-2-carbonitrile | |
15111480 | |
95% |
1452-16-0 | |
MFCD09702472 | |
UN3439 | |
Slightly soluble in water. | |
C1=CSC(=N1)C#N | |
110.134 | |
110.14 | |
Solid |
Chemical Identifiers
1452-16-0 | |
110.134 | |
ZIRGWUZHKJDYKT-UHFFFAOYSA-N | |
15111480 | |
C1=CSC(=N1)C#N |
C4H2N2S | |
MFCD09702472 | |
2-cyanothiazole, thiazole-2-carbonitrile, 2-thiazolecarbonitrile, 2-cyanothiazol | |
1,3-thiazole-2-carbonitrile |
Safety and Handling
GHS H Statement
H371-H302-H312-H332-H315-H319
May cause damage to organs.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P309+P311-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H371
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only