missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyanopyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5468903

 View more versions of this product

Catalog No. AAH5468903



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is used in suzuki reactions.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents, air, heat. Refrigerated (2-4°C).
Specifications

Specifications

2-Cyanopyridine-5-boronic acid pinacol ester
1g
C12H15BN2O2
2-cyano pyridine-5-boronic acid pinacol ester
IXTBQKLZPOYJFJ-UHFFFAOYSA-N
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile
16414188
96%
105°C to 106°C
741709-63-7
MFCD06657825
Slightly soluble in water.
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C#N
230.074
230.07

Chemical Identifiers

741709-63-7
230.074
IXTBQKLZPOYJFJ-UHFFFAOYSA-N
16414188
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C#N
C12H15BN2O2
MFCD06657825
2-cyano pyridine-5-boronic acid pinacol ester
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents