Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyanopyridine-3-boronic acid pinacol ester, 95%

$745.85 - $745.85

Chemical Identifiers

Chemical Name or Material 2-Cyanopyridine-3-boronic acid pinacol ester
CAS 878194-93-5
Molecular Formula C12H15BN2O2
Formula Weight 230.07
MDL Number MFCD08458477
Synonym 2-Cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2733203

  SDS
alfa aesar™
H2733203
1g Each for $745.85
Description & Specifications

Specifications

Formula Weight 230.07
Quantity 1g
MDL Number MFCD08458477
Chemical Name or Material 2-Cyanopyridine-3-boronic acid pinacol ester
CAS 878194-93-5
Molecular Formula C12H15BN2O2
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 58° to 61°C
Synonym 2-Cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Danger
Assay 95%

Applications
A pyridylborane derivative evaluated for its kinase inhibitory activity. Also used in immunodiagnostics. Used as intermediate. As syntheses material intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.