missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyanophenol, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 161411000

 View more versions of this product

Catalog No. AC161411000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Cyanophenol
Glass bottle
White to Brown
100g
611-20-1
100.0
C7H5NO
MFCD00002145
2-cyanophenol
CHZCERSEMVWNHL-UHFFFAOYSA-N
2-hydroxybenzonitrile
11907
Crystalline Powder
Authentic
149.0°C (14.0 mmHg)
95.0°C to 98.0°C
99%
98.5
99%
NCC6H4OH
10, 96
Solubility in water: soluble
C1=CC=C(C(=C1)C#N)O
119.12
119.12
99%

Chemical Identifiers

611-20-1
119.12
CHZCERSEMVWNHL-UHFFFAOYSA-N
11907
C1=CC=C(C(=C1)C#N)O
C7H5NO
MFCD00002145
2-cyanophenol
2-hydroxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Danger

EINECSNumber : 210-259-3

TSCA : TSCA

SDS Documents