missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Cyanomethyl)benzeneboronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H53255MD

 View more versions of this product

Catalog No. AAH53255MD



Description

Description

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents
Specifications

Specifications

2-(Cyanomethyl)benzeneboronic acid pinacol ester
325141-71-7
MFCD02179457
FIARMBRSPSECPP-UHFFFAOYSA-N
2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
3832884
97%
250mg
C14H18BNO2
2-cyanomethylphenylboronic acid, pinacol ester, 2-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-cyanomethylphenyl boronic acid, pinacol ester, 2-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, benzeneacetonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-cyanomethylphenyl boronic acid,pinacol ester, 2-cyanomethyl benzeneboronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl acetonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CC#N
243.113
243.11

Chemical Identifiers

325141-71-7
243.113
FIARMBRSPSECPP-UHFFFAOYSA-N
3832884
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CC#N
C14H18BNO2
MFCD02179457
2-cyanomethylphenylboronic acid, pinacol ester, 2-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-cyanomethylphenyl boronic acid, pinacol ester, 2-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, benzeneacetonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-cyanomethylphenyl boronic acid,pinacol ester, 2-cyanomethyl benzeneboronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl acetonitrile
2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents