missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyanoethylhydrazine, 98%, ACROS Organics™

$40.24 - $40.24

Chemical Identifiers

CAS 353-07-1
Molecular Formula C3H7N3
Molecular Weight (g/mol) 85.11
MDL Number MFCD00043620
InChI Key VIHRIIARIFUQLC-UHFFFAOYSA-N
Synonym 2-cyanoethylhydrazine, beta-cyanoethylhydrazine, 2-cyanoethyl hydrazine, cyanoethylhydrazine, unii-l6h109c98b, 2-cyanoethyl hydrazine, 3-hydrazinopropionitrile, propanenitrile, 3-hydrazino, 2-cyanoethylhydrazine, 3-hydrazinopropanenitrile
PubChem CID 136177
IUPAC Name 3-hydrazinylpropanenitrile
SMILES C(CNN)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC335480050 SDS View Documents Acros Organics
335480050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC335480010 SDS View Documents Acros Organics
335480010
1g Glass bottle Each for $40.24
Specifications

Chemical Identifiers

353-07-1
85.11
VIHRIIARIFUQLC-UHFFFAOYSA-N
136177
C(CNN)C#N
C3H7N3
MFCD00043620
2-cyanoethylhydrazine, beta-cyanoethylhydrazine, 2-cyanoethyl hydrazine, cyanoethylhydrazine, unii-l6h109c98b, 2-cyanoethyl hydrazine, 3-hydrazinopropionitrile, propanenitrile, 3-hydrazino, 2-cyanoethylhydrazine, 3-hydrazinopropanenitrile
3-hydrazinylpropanenitrile

Specifications

Authentic
1.4740 to 1.4780 (20°C, 589nm)
5g
97.5
98%
NH2NHCH2CH2CN
2-cyanoethylhydrazine, beta-cyanoethylhydrazine, 2-cyanoethyl hydrazine, cyanoethylhydrazine, unii-l6h109c98b, 2-cyanoethyl hydrazine, 3-hydrazinopropionitrile, propanenitrile, 3-hydrazino, 2-cyanoethylhydrazine, 3-hydrazinopropanenitrile
C(CNN)C#N
85.11
85.1
≥97.5%
Glass bottle
Yellow
353-07-1
100.0
C3H7N3
MFCD00043620
VIHRIIARIFUQLC-UHFFFAOYSA-N
3-hydrazinylpropanenitrile
136177
Liquid
2-Cyanoethylhydrazine, 98%
Safety and Handling

Safety and Handling

GHS P Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

TSCA : TSCA

Recommended Storage : product darkens on exposure to air