Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyanoethylhydrazine, 95%

Chemical Identifiers

Chemical Name or Material 2-Cyanoethylhydrazine
CAS 353-07-1
Molecular Formula C3H7N3
Linear Formula NCCH2CH2NHNH2
Formula Weight 85.11
MDL Number MFCD00043620
UN Number UN2927
Beilstein 773834
Synonym 3-Hydrazinopropionitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2522706

  SDS
alfa aesar™
B2522706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2522703

  SDS
alfa aesar™
B2522703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2522714

  SDS
alfa aesar™
B2522714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 85.11
Chemical Name or Material 2-Cyanoethylhydrazine
Assay 95%
Molecular Formula C3H7N3
Linear Formula NCCH2CH2NHNH2
MDL Number MFCD00043620
CAS 353-07-1
UN Number UN2927
Beilstein 773834
Synonym 3-Hydrazinopropionitrile
Health Hazard 1 Danger
Boiling Point 76° to 79°C (0.5mmHg)
Health Hazard 2 GHS H Statement
H300-H314
Fatal if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
Flash Point 109°C (228°F)
Quantity 5g

Applications
2-Cyanoethylhydrazine is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H300-H314
Fatal if swallowed.
Causes severe skin burns and eye damage.

2° to 8°C