Learn More
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 379740010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite | |
0.9490g/mL | |
60°C | |
Glass bottle | |
1.47 | |
0.949 | |
RKVHNYJPIXOHRW-UHFFFAOYSA-N | |
3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile | |
128153 | |
95% |
102691-36-1 | |
98.0°C to 100.0°C (5.0 mmHg) | |
95% | |
C15H32N3OP | |
1 g | |
3-bis diisopropylamino phosphino oxy propanenitrile, bis diisopropylamino 2-cyanoethoxy phosphine, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite, phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite, 2-cyanoethyl tetraisopropylphosphorodiamidite, cetipd, 3-bis diisopropylamino phosphinooxy propanenitrile | |
CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N | |
301.41 | |
301.41 |
Chemical Identifiers
102691-36-1 | |
301.41 | |
3-bis diisopropylamino phosphino oxy propanenitrile, bis diisopropylamino 2-cyanoethoxy phosphine, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite, phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite, 2-cyanoethyl tetraisopropylphosphorodiamidite, cetipd, 3-bis diisopropylamino phosphinooxy propanenitrile | |
3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile |
C15H32N3OP | |
RKVHNYJPIXOHRW-UHFFFAOYSA-N | |
128153 | |
CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surface
GHS Signal Word: Danger
RUO – Research Use Only