Please login to your online account to display your discounted pricing

2-Cyanobenzene-1-sulfonyl chloride, ≥95%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  TL00431DA

 View more versions of this product

Catalog No. TL00431DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 201.63
Quantity 1g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
CAS 69360-26-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 2-Cyanobenzene-1-sulfonyl chloride
Packaging Amber glass bottle
Molecular Formula C7H4ClNO2S
MDL Number MFCD00052341
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Cyanobenzene-1-sulfonyl chloride
CAS 69360-26-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H4ClNO2S
Formula Weight 201.63
MDL Number MFCD00052341
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents