Please login to your online account to display your discounted pricing

2-Cyanobenzene-1-sulfonyl chloride, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Cyanobenzene-1-sulfonyl chloride
CAS 69360-26-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H4ClNO2S
Formula Weight 201.63
MDL Number MFCD00052341
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00431DA

  SDS
fisher scientific company
TL00431DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.63
Quantity 1g
Assay 95%
CAS Min % 95.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS 69360-26-5
Assay Percent Range 95%
Molecular Formula C7H4ClNO2S
Chemical Name or Material 2-Cyanobenzene-1-sulfonyl chloride
MDL Number MFCD00052341
Packaging Amber glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.