Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyanobenzamide, 98%

$39.03 - $145.70

Chemical Identifiers

Chemical Name or Material 2-Cyanobenzamide
CAS 17174-98-0
Molecular Formula C8H6N2O
Formula Weight 146.15
MDL Number MFCD00014797
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2159603

  SDS
alfa aesar™
B2159603
1g Each for $39.03

AAB2159606

  SDS
alfa aesar™
B2159606
5g Each for $145.70
Description & Specifications

Specifications

Formula Weight 146.15
TSCA TSCA
Chemical Name or Material 2-Cyanobenzamide
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
CAS 17174-98-0
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 1 Danger
MDL Number MFCD00014797
Molecular Formula C8H6N2O
Melting Point 172° to 173°C
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.