missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyanobenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC371860250

 View more versions of this product

Catalog No. AC371860250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Powder, Crystals and/or Chunks
131.13
White to orange to beige
100.0
Glass bottle
7468-67-9
98%
MFCD00017503
≥97.5%
25g
102° to 106°C
NCC6H4CHO
C8H5NO
2-Cyanobenzaldehyde
97.5
Authentic

Chemical Identifiers

7468-67-9
100.0
C8H5NO
131.13
2-Cyanobenzaldehyde
97.5
98%
NCC6H4CHO
MFCD00017503
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning