Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyanobenzaldehyde, 98%

$69.00 - $207.93

Chemical Identifiers

Chemical Name or Material 2-Cyanobenzaldehyde
CAS 7468-67-9
Molecular Formula C8H5NO
Formula Weight 131.13
MDL Number MFCD00017503
Synonym 2-Formylbenzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2902706

  SDS
alfa aesar™
H2902706
5g Each for $207.93

AAH2902703

  SDS
alfa aesar™
H2902703
1g Each for $69.00
Description & Specifications

Specifications

Formula Weight 131.13
Quantity 5g
Assay 98%
Melting Point 102° to 106°C
Odor Characteristic
MDL Number MFCD00017503
Chemical Name or Material 2-Cyanobenzaldehyde
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 7468-67-9
Synonym 2-Formylbenzonitrile
Health Hazard 1 Warning
Molecular Formula C8H5NO
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.