missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyano-N,N-diethylacetamide, TCI America™

Manufacturer:  TCI America C282225G

 View more versions of this product

Catalog No. C282225G



Specifications

Specifications

2-Cyano-N,N-diethylacetamide
Yellow
26391-06-0
MFCD01332383
CCN(CC)C(=O)CC#N
140.186
CHEBI:48386
Clear Liquid at 20°C
114°C
25g
C7H12N2O
RYSHIRFTLKZVIH-UHFFFAOYSA-N
2-cyano-N,N-diethylacetamide
3854515
140.19
≥98.0% (GC)

Chemical Identifiers

26391-06-0
140.186
RYSHIRFTLKZVIH-UHFFFAOYSA-N
CHEBI:48386
CCN(CC)C(=O)CC#N
C7H12N2O
MFCD01332383
3854515
2-cyano-N,N-diethylacetamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents