Learn More
2-Cyano-6-methylpyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 425920010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Cyano-6-methylpyridine | |
96% min. (GC) | |
C7H6N2 | |
1g | |
CMADFEQMYFNYCF-UHFFFAOYSA-N | |
6-methylpyridine-2-carbonitrile | |
74182 | |
97% |
1620-75-3 | |
Glass bottle | |
MFCD00190586 | |
6-methylpicolinonitrile, 2-cyano-6-methylpyridine, 6-methyl-2-pyridinecarbonitrile, 2-methyl-6-cyanopyridine, 6-methyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 6-methyl, 6-methyl-2-cyanopyridine, 2-cyano-6-methyl pyridine, 2-cyano-6-methylpyridine 2-cyano-6-picoline, 6-methylpyridin-2-carbonitril | |
CC1=CC=CC(=N1)C#N | |
118.14 | |
118.14 |
Chemical Identifiers
1620-75-3 | |
118.14 | |
CMADFEQMYFNYCF-UHFFFAOYSA-N | |
74182 | |
CC1=CC=CC(=N1)C#N |
C7H6N2 | |
MFCD00190586 | |
6-methylpicolinonitrile, 2-cyano-6-methylpyridine, 6-methyl-2-pyridinecarbonitrile, 2-methyl-6-cyanopyridine, 6-methyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 6-methyl, 6-methyl-2-cyanopyridine, 2-cyano-6-methyl pyridine, 2-cyano-6-methylpyridine 2-cyano-6-picoline, 6-methylpyridin-2-carbonitril | |
6-methylpyridine-2-carbonitrile |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 216-588-9
RUO â Research Use Only