Please login to your online account to display your discounted pricing

2-Cyano-6-methoxybenzothiazole, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Cyano-6-methoxybenzothiazole
CAS 943-03-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H6N2OS
Formula Weight 190.23
MDL Number MFCD00010537
Synonym 6-Methoxy-2-benzothiazolecarbonitrile
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC225362500

  SDS
thermo fisher scientific
AC225362500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC225360010

  SDS
thermo fisher scientific
225360010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 190.23
Quantity 250mg
Color Off-white to light yellow
Melting Point 128° to 131°C
CAS Max % 100.0
Chemical Name or Material 2-Cyano-6-methoxybenzothiazole
Assay Percent Range 99%
Molecular Formula C9H6N2OS
MDL Number MFCD00010537
CAS 943-03-3
Synonym 6-Methoxy-2-benzothiazolecarbonitrile
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.